EMAIL THIS PAGE TO A FRIEND
Prostaglandins, leukotrienes, and essential fatty acids
Docosahexaenoic acid supplementation in lactating women increases breast milk and plasma docosahexaenoic acid concentrations and alters infant omega 6:3 fatty acid ratio.
PMID 25701002
Abstract
This study investigated the effects of docosahexaenoic acid (DHA) supplementation on the fatty acid composition of breast milk and plasma concentrations in lactating women and their infants. Eighty-nine lactating women 4-6 weeks post-partum received placebo, 200 mg or 400 mg DHA for 6 weeks with usual diets. Breast milk fatty acids and maternal plasma fatty acids were measured at the beginning and end of the study and infant plasma at the end of the study. Breast milk and maternal plasma DHA were significantly greater with 200 mg and 400 mg DHA compared with placebo (50% and 123% breast milk p<0.05; 71% and 101% plasma, p<0.0001), respectively. Infant plasma omega 6:3 and arachidonic acid (AA):DHA were significantly greater in the placebo group compared to both supplement groups (67% and 106%; 71% and 116%, respectively, p<0.05). DHA supplementation impacts infant fatty acids important for brain development and breast milk fatty acid composition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
08339
5-Aminolevulinic acid hydrochloride, ≥97.0% (AT)
C5H9NO3 · HCl