EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Impact of deoxynivalenol (DON) contaminated feed on intestinal integrity and immune response in swine.
PMID 25701311
Abstract
This study was performed to characterize the influence of consuming DON naturally contaminated feeds on pig's intestinal immune defenses, antibody response and cellular immunity. Sixteen 4-week-old piglets were randomly allocated to two dietary treatments: control diet or diet contaminated with 3.5 mg DON/kg. At days 7 and 21, animals were immunized with ovalbumin (OVA). On day 42, intestinal samples were collected for measurement of gene expression involved in immune response, oxidative status and barrier function. Primary IgG antibody response to OVA was increased in pigs fed DON diet compared to control animals. In the ileum of pigs fed DON diet, claudin, occludin, and vimentin genes involved in integrity and barrier function were down-regulated compared to controls. Results also revealed that expression of two chemokines (IL-8, CXCL10), interferon-γ, and major antioxidant glutathione peroxidase 2 (GPX-2) were up-regulated whereas expression of genes encoding enzymatic antioxidants including GPX-3, GPX-4 and superoxide dismutase 3 (SOD-3) were down-regulated in pigs fed DON-contaminated diet. These results strongly suggest that ingestion of DON naturally contaminated feed impaired intestinal barrier and immunological functions by modulating expression of genes coding for proteins involved in tight junctions, tissue remodelling, inflammatory reaction, oxidative stress reaction and immune response.