EMAIL THIS PAGE TO A FRIEND
Food chemistry
Multiresidue method for the determination of pharmacologically active substances in egg and honey using a continuous solid-phase extraction system and gas chromatography-mass spectrometry.
PMID 25704684
Abstract
A sensitive, selective, efficient gas chromatography-mass spectrometry method for the simultaneous determination of 22 pharmacologically active substances (antibacterials, nonsteroidal antiinflammatories, antiseptics, antiepileptics, lipid regulators, β-blockers and hormones) in eggs and honey was developed. The sample pretreatment includes precipitation of proteins and lipids with acetonitrile:water (3:2, v/v), centrifugation and continuous solid-phase extraction for cleanup and preconcentration. The proposed method was validated with quite good analytical results including limits of detection of 0.4-3.3 ng/kg for 2g of sample and good linearity (r(2)>0.995) throughout the studied concentration ranges. The recoveries of analytes from real honey and egg samples spiked at concentrations of 15-2,000 ng/kg fell in the range 87-102%, with relative standard deviations from 2.6% to 7.0%. The method was successfully used to determine the target compounds in various types of eggs (hen, quail and duck) and honey samples (flower, forest, acacia, sunflower, clover and pine tree). Two samples of hen eggs bought at supermarkets and one of quail eggs were found to contain florfenicol, pyrimethamine, estrone and 17β-estradiol at levels from 0.095 to 2.7 μg/kg.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284785
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
C14H14O3
524190
5-Chloro-2-(2,4-dichlorophenoxy)phenol, 97%
C12H7Cl3O2
94496
Carbamazepine, analytical standard
C15H12N2O
C0450000
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
C15H12N2O
1093001
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
C15H12N2O
PHR1067
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
C15H12N2O
92360
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
C3H9ClSi
89595
Chlorotrimethylsilane, for GC derivatization, ≥99.0% (GC)
C3H9ClSi
E9750
Estrone, ≥99%
C18H22O2
E1274
Estrone, meets USP testing specifications
C18H22O2
46573
Estrone, VETRANAL, analytical standard
C18H22O2
1255001
Estrone, United States Pharmacopeia (USP) Reference Standard
C18H22O2
E1700000
Estrone, European Pharmacopoeia (EP) Reference Standard
C18H22O2
PHR1535
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material
C18H22O2
F1427
Florfenicol, analytical standard, for drug analysis
C12H14Cl2FNO4S
33586
Flunixin, VETRANAL, analytical standard
C14H11F3N2O2
72779
Irgasan, ≥97.0% (HPLC)
C12H7Cl3O2
K1751
Ketoprofen, ≥98% (TLC)
C16H14O3
K2012
Ketoprofen, meets USP testing specifications
C16H14O3
1356632
Ketoprofen, United States Pharmacopeia (USP) Reference Standard
C16H14O3
K2000000
Ketoprofen, European Pharmacopoeia (EP) Reference Standard
C16H14O3
34016
Ketoprofen, VETRANAL, analytical standard
C16H14O3
PHR1375
Ketoprofen, Pharmaceutical Secondary Standard; Certified Reference Material
C16H14O3
M4267
Mefenamic acid
C15H15NO2
92574
Mefenamic acid, analytical standard
C15H15NO2
M0252600
Mefenamic acid, European Pharmacopoeia (EP) Reference Standard
C15H15NO2
34148
Metolachlor OA, PESTANAL®, analytical standard
C15H21NO4
15222
N,O-Bis(trimethylsilyl)trifluoroacetamide, for GC derivatization, ≥99.0%
C8H18F3NOSi2
15238
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, contains 1% TMCS, 99% (excluding TMCS)
C8H18F3NOSi2
15209
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, contains 10% TMCS, 98% (excluding TMCS)
C8H18F3NOSi2