PloS one
Effect of preoperatively continued aspirin use on early and mid-term outcomes in off-pump coronary bypass surgery: a propensity score-matched study of 1418 patients.
PMID 25706957
Abstract
To date, effect of preoperatively continued aspirin administration in off-pump coronary artery bypass grafting (CABG) is less known. We aimed to assess the effect of preoperatively continued aspirin use on early and mid-term outcomes in patients receiving off-pump CABG. From October 2009 to September 2013 at the Fuwai Hospital, 709 preoperative aspirin users were matched with unique 709 nonaspirin users using propensity score matching to obtain risk-adjusted outcome comparisons between the two groups. Early outcomes were in-hospital death, stroke, intra- and post-operative blood loss, reoperation for bleeding and blood product transfusion. Major adverse cardiac events (death, myocardial infarction or repeat revascularization), angina recurrence and cardiogenic readmission were considered as mid-term endpoints. There were no significant differences among the groups in baseline characteristics after propensity score matching. The median intraoperative blood loss (600 ml versus 450 ml, P = 0.56), median postoperative blood loss (800 ml versus 790 ml, P = 0.60), blood transfusion requirements (25.1% versus 24.4%, P = 0.76) and composite outcome of in-hospital death, stroke and reoperation for bleeding (2.8% versus 1.6%, P = 0.10) were similar in aspirin and nonaspirin use group. At about 4 years follow-up, no significant difference was observed among the aspirin and nonaspirin use group in major adverse cardiac events free survival estimates (95.7% versus 91.5%, P = 0.23) and freedom from cardiogenic readmission (88.5% versus 85.3%, P = 0.77) whereas the angina recurrence free survival rates was 83.7% and 73.9% in the aspirin and nonaspirin use group respectively (P = 0.02), with odd ratio for preoperative aspirin estimated at 0.71 (95% confidence interval, 0.49-1.04, P = 0.08). Preoperatively continued aspirin use was not associated with increased risk of intra- and post-operative blood loss, blood transfusion requirements and composite outcome of in-hospital death, stroke and reoperation for bleeding in off-pump CABG. Preoperative aspirin use tended to decrease the hazard of mid-term angina recurrence.