PloS one
Role of exopolysaccharide in Aggregatibacter actinomycetemcomitans-induced bone resorption in a rat model for periodontal disease.
PMID 25706999
Abstract
Aggregatibacter actinomycetemcomitans a causative agent of periodontal disease in humans, forms biofilm on biotic and abiotic surfaces. A. actinomycetemcomitans biofilm is heterogeneous in nature and is composed of proteins, extracellular DNA and exopolysaccharide. To explore the role played by the exopolysaccharide in the colonization and disease progression, we employed genetic reduction approach using our rat model of A. actinomycetemcomitans-induced periodontitis. To this end, a genetically modified strain of A. actinomycetemcomitans lacking the pga operon was compared with the wild-type strain in the rat infection model. The parent and mutant strains were primarily evaluated for bone resorption and disease. Our study showed that colonization, bone resorption/disease and antibody response were all elevated in the wild-type fed rats. The bone resorption/disease caused by the pga mutant strain, lacking the exopolysaccharide, was significantly less (P < 0.05) than the bone resorption/disease caused by the wild-type strain. Further analysis of the expression levels of selected virulence genes through RT-PCR showed that the decrease in colonization, bone resorption and antibody titer in the absence of the exopolysaccharide might be due to attenuated levels of colonization genes, flp-1, apiA and aae in the mutant strain. This study demonstrates that the effect exerted by the exopolysaccharide in A. actinomycetemcomitans-induced bone resorption has hitherto not been recognized and underscores the role played by the exopolysaccharide in A. actinomycetemcomitans-induced disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9346
Liquified Phenol, meets USP testing specifications, ≥89.0%
C6H6O
P1037
Phenol, for molecular biology
C6H6O
P5566
Phenol, BioXtra, ≥99.5% (GC)
C6H6O
328111
Phenol, unstabilized, purified by redistillation, ≥99%
C6H6O
242322
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
C6H6O
77607
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
C6H6O
77608
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
C6H6O
77613
Phenol, for molecular biology, ~90% (T), liquid
C6H6O
35952
Phenol, PESTANAL®, analytical standard
C6H6O
33517
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
C6H6O
16016
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
C6H6O
16017
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
C6H6O
W322318
Phenol, ≥99%
C6H6O
PHR1047
Phenol
C6H6O
W322340
Phenol, natural, 97%, FG
C6H6O
1524806
Phenol, United States Pharmacopeia (USP) Reference Standard
C6H6O
P4557
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
C6H6O
P4682
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
C6H6O
46344
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
C6H6O
48688
Phenol solution, certified reference material, 500 μg/mL in methanol
C6H6O
40063
Phenol solution, 5000 μg/mL in methanol, certified reference material
C6H6O
791741
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
C2H3NaO2
S2889
Sodium acetate, anhydrous, for molecular biology, ≥99%
C2H3NaO2
S3272
Sodium acetate, for electrophoresis, ≥99%, crystalline
C2H3NaO2
S5636
Sodium acetate, powder, BioReagent, for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
C2H3NaO2
S1429
Sodium acetate, meets USP testing specifications, anhydrous
C2H3NaO2
1613407
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
C2H3NaO2
229873
Sodium acetate, 99.995% trace metals basis
C2H3NaO2
S7545
Sodium acetate, BioXtra, ≥99.0%
C2H3NaO2
110191
Sodium acetate, ReagentPlus®, 99%
C2H3NaO2