EMAIL THIS PAGE TO A FRIEND
British journal of haematology
Ricolinostat (ACY-1215) induced inhibition of aggresome formation accelerates carfilzomib-induced multiple myeloma cell death.
PMID 25709080
Abstract
Proteasome inhibition induces the accumulation of aggregated misfolded/ubiquitinated proteins in the aggresome; conversely, histone deacetylase 6 (HDAC6) inhibition blocks aggresome formation. Although this rationale has been the basis of proteasome inhibitor (PI) and HDAC6 inhibitor combination studies, the role of disruption of aggresome formation by HDAC6 inhibition has not yet been studied in multiple myeloma (MM). The present study aimed to evaluate the impact of carfilzomib (CFZ) in combination with a selective HDAC6 inhibitor (ricolinostat) in MM cells with respect to the aggresome-proteolysis pathway. We observed that combination treatment of CFZ with ricolinostat triggered synergistic anti-MM effects, even in bortezomib-resistant cells. Immunofluorescent staining showed that CFZ increased the accumulation of ubiquitinated proteins and protein aggregates in the cytoplasm, as well as the engulfment of aggregated ubiquitinated proteins by autophagosomes, which was blocked by ricolinostat. Electron microscopy imaging showed increased autophagy triggered by CFZ, which was inhibited by the addition of ACY-1215. Finally, an in vivo mouse xenograft study confirmed a decrease in tumour volume, associated with apoptosis, following treatment with CFZ in combination with ricolinostat. Our results suggest that ricolinostat inhibits aggresome formation, caused by CFZ-induced inhibition of the proteasome pathway, resulting in enhanced apoptosis in MM cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
220051
N-Iodosuccinimide, 95%
C4H4INO2