EMAIL THIS PAGE TO A FRIEND
BioMed research international
Exercise-induced neuroprotection in the spastic Han Wistar rat: the possible role of brain-derived neurotrophic factor.
PMID 25710032
Abstract
Moderate aerobic exercise has been shown to enhance motor skills and protect the nervous system from neurodegenerative diseases, like ataxia. Our lab uses the spastic Han Wistar rat as a model of ataxia. Mutant rats develop forelimb tremor and hind limb rigidity and have a decreased lifespan. Our lab has shown that exercise reduced Purkinje cell degeneration and delayed motor dysfunction, significantly increasing lifespan. Our study investigated how moderate exercise may mediate neuroprotection by analyzing brain-derived neurotrophic factor (BDNF) and its receptor TrkB. To link BDNF to exercise-induced neuroprotection, mutant and normal rats were infused with the TrkB antagonist K252a or vehicle into the third ventricle. During infusion, rats were subjected to moderate exercise regimens on a treadmill. Exercised mutants receiving K252a exhibited a 21.4% loss in Purkinje cells compared to their controls. Cerebellar TrkB expression was evaluated using non-drug-treated mutants subjected to various treadmill running regimens. Running animals expressed three times more TrkB than sedentary animals. BDNF was quantified via Sandwich ELISA, and cerebellar expression was found to be 26.6% greater in mutant rats on 7-day treadmill exercise regimen compared to 30 days of treadmill exercise. These results suggest that BDNF is involved in mediating exercise-induced neuroprotection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03378
Chloride atomic spectroscopy standard concentrate 10.00 g Cl-, 10.00 g/L, for 1 l standard solution, analytical standard
HCl
94563
Dimethyl sulfoxide, analytical standard
C2H6OS
01934
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
C2H6OS
1211006
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
C2H6OS
D2050000
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
C2H6OS
84426
Hydrochloric acid, purum p.a., fuming 37%, ≥37% (T)
HCl
17933
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization
HCl
17934
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization
HCl
17935
Hydrogen chloride – methanol solution, ~1.25 M HCl, for GC derivatization
HCl
S8032
Sodium azide, BioXtra
N3Na
438456
Sodium azide, ≥99.99% trace metals basis
N3Na
71289
Sodium azide, BioUltra, ≥99.5% (T)
N3Na
71290
Sodium azide, purum p.a., ≥99.0% (T)
N3Na
71383
Sodium chloride, tested according to Ph.Eur.
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
ClNa
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa
71177
Sodium standard for AAS, ready-to-use, traceable to BAM, in H2O
ClNa