EMAIL THIS PAGE TO A FRIEND
International journal of dermatology
HIPK2 expression in progression of cutaneous epithelial neoplasm.
PMID 25711204
Abstract
Homeodomain-interacting protein kinase 2 (HIPK2) is responsible for a DNA damage response, centrally regulating p53. The aberrant HIPK2 expression is known to be involved in carcinogenesis in several malignancies. However, the correlation of HIPK2 expression along with progression of cutaneous epithelial neoplasm has not been investigated. Using immunohistochemistry and real-time reverse transcription-polymerase chain reaction, we examined the correlation between HIPK2 and HIPK2-related protein expressions and the progression of some cutaneous epithelial neoplasms (i.e., actinic keratosis, Bowen's disease, keratoacanthoma, squamous cell carcinoma, and basal cell carcinoma). HIPK2 expression was distinct between preinvasive and invasive lesions: the expression decreased in keratoacanthoma (none of eight) and squamous cell carcinoma (five of 35) compared to actinic keratosis (12 of 19) and Bowen's disease (10 of 23) (Pxa0