EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
AMP-regulated protein kinase activity in the hearts of mice treated with low- or high-fat diet measured using novel LC-MS method.
PMID 25711403
Abstract
AMP-regulated protein kinase (AMPK) is involved in regulation of energy-generating pathways in response to the metabolic needs in different organs including the heart. The activity of AMPK is mainly controlled by AMP concentration that in turn could be affected by nucleotide metabolic pathways. This study aimed to develop a procedure for measurement of AMPK activity together with nucleotide metabolic enzymes and its application for studies of mice treated with high-fat diet. The method developed was based on analysis of conversion of AMARA peptide to pAMARA by partially purified heart homogenate by liquid chromatography/mass spectrometry (LC/MS). Activities of the enzymes of nucleotide metabolism were evaluated by analysis of conversion of substrates into products by HPLC. The method was applied for analysis of hearts of mice fed 12 weeks with low- (LFD) or high-fat diet (HFD). The optimized method for AMPK activity analysis (measured in presence of AMP) revealed change of activity from 0.089 ± 0.035 pmol/min/mg protein in LFD to 0.024 ± 0.002 in HFD. This coincided with increase of adenosine deaminase (ADA) activity from 0.11 ± 0.02 to 0.19 ± 0.06 nmol/mg tissue/min and decrease of AMP-deaminase (AMPD) activity from 1.26 ± 0.35 to 0.56 ± 0.15 nmol/mg tissue/min for LFD and HFD, respectively. We have proven quality of our LC/MS method for analysis of AMPK activity. We observed decrease in AMPK activity in the heart of mice treated with high-fat diet. However, physiological consequences of this change could be modulated by decrease in AMPD activity.