EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Interim anatase coating layer stabilizes rutile@Crx Oy photoanode for visible-light-driven water oxidation.
PMID 25712145
Abstract
Ternary core-shell heterostructured rutile@anatase@Crx Oy nanorod arrays were elaborately designed as photoanodes for efficient photoelectrochemical water splitting under visible-light illumination. The four-fold enhanced and stabilized visible-light photocurrent highlights the unique role of the interim anatase layer in accelerating the interfacial charge transfer from the Crx Oy chromophore to rutile nanorods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
244112
Titanium(IV) butoxide, reagent grade, 97%
C16H36O4Ti