EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Complete biosynthetic pathway of the C50 carotenoid bacterioruberin from lycopene in the extremely halophilic archaeon Haloarcula japonica.
PMID 25712483
Abstract
Haloarcula japonica, an extremely halophilic archaeon that requires high concentrations of NaCl for growth, accumulates the C50 carotenoid bacterioruberin (BR). By homology analysis, a gene cluster, including c0507, c0506, and c0505, was found and predicted to be involved in the synthesis of bacterioruberin. To elucidate the function of the encoded enzymes, we constructed Ha. japonica mutants of these genes and analyzed carotenoids produced by the mutants. Our research showed that c0507, c0506, and c0505 encoded a carotenoid 3,4-desaturase (CrtD), a bifunctional lycopene elongase and 1,2-hydratase (LyeJ), and a C50 carotenoid 2",3"-hydratase (CruF), respectively. The above three carotenoid biosynthetic enzymes catalyze the reactions that convert lycopene to bacterioruberin in Ha. japonica. This is the first identification of functional CrtD and CruF in archaea and elucidation of the complete biosynthetic pathway of bacterioruberin from lycopene. Haloarcula japonica, an extremely halophilic archaeon, accumulates the C50 carotenoid bacterioruberin (BR). In this study, we have identified three BR biosynthetic enzymes and have elucidated their functions. Among them, two enzymes were found in an archaeon for the first time. Our results revealed the biosynthetic pathway responsible for production of BR in Ha. japonica and provide a basis for investigating carotenoid biosynthetic pathways in other extremely halophilic archaea. Elucidation of the carotenoid biosynthetic pathway in Ha. japonica may also prove useful for producing the C50 carotenoid BR efficiently by employing genetically modified haloarchaeal strains.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73148
3-Nitrobenzyl alcohol, for mass spectrometry, ≥99.5%
C7H7NO3
146056
3-Nitrobenzyl alcohol, 98%
C7H7NO3
266167
Manganese, chips, thickness <2.0 mm, 99%
Mn
266132
Manganese, powder, −325 mesh, ≥99% trace metals basis
Mn
463728
Manganese, powder, ≥99.9% trace metals basis
Mn
GF74177736
Manganese, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF99005193
Manganese, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 98.7%
Mn
GF90559794
Manganese, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 98.7%
Mn
GF03428625
Manganese, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
Mn
GF31519975
Manganese, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
Mn
GF60677646
Manganese, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 98.7%
Mn
GF64815239
Manganese, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 98.7%
Mn
GF41418378
Manganese, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 98.7%
Mn
GF33640860
Manganese, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
Mn
GF85367605
Manganese, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
Mn
GF87981564
Manganese, foil, not light tested, 50x50mm, thickness 0.006mm, permanent polyester support, 98.7%
Mn
GF53022833
Manganese, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%
Mn
GF71274258
Manganese, foil, not light tested, 50x50mm, thickness 0.008mm, permanent polyester support, 98.7%
Mn
GF70055524
Manganese, foil, not light tested, 50x50mm, thickness 0.009mm, permanent polyester support, 98.7%
Mn
GF86610539
Manganese, foil, not light tested, 50x50mm, thickness 0.0125mm, permanent polyester support, 98.7%
Mn
GF00832081
Manganese, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%
Mn
GF61911288
Manganese, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF94645233
Manganese, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 98.7%
Mn
GF05710430
Manganese, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 98.7%
Mn
GF27632055
Manganese, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 98.7%
Mn
GF11021094
Manganese, rod, 10mm, diameter 4.0mm, cast, 99.5%
Mn
GF34573697
Manganese, rod, 35mm, diameter 2.0mm, cast, 99.5%
Mn
GF38495885
Manganese, rod, 50mm, diameter 4.0mm, cast, 99.5%
Mn
GF42039918
Manganese, rod, 50mm, diameter 6.35mm, cast, 99.5%
Mn
GF83932768
Manganese, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Mn