EMAIL THIS PAGE TO A FRIEND
Journal of molecular and cellular cardiology
A crosstalk between chromatin remodeling and histone H3K4 methyltransferase complexes in endothelial cells regulates angiotensin II-induced cardiac hypertrophy.
PMID 25712920
Abstract
Angiotensin II (Ang II) induces cardiac hypertrophy and fibrosis in part by stimulating endothelin (ET-1) transcription. The involvement of the epigenetic machinery in this process is largely undefined. In the present study, we examined the epigenetic maneuvering underlying cardiac hypertrophy and fibrosis following ET-1 transactivation by Ang II. In response to Ang II stimulation, core components of the mammalian chromatin remodeling complex (Brahma-related gene 1, or Brg1, and Brahma or Brm) and histone H3K4 methylation complex (Ash2, absent, small, or homeotic discs 2, or Ash2 and WD domain repeat 5, or Wdr5) were recruited to the ET-1 promoter region in endothelial cells. Over-expression of Brg1/Brm or Ash2/Wdr5 enhanced while depletion of Brg1/Brm or Ash2/Wdr5 attenuated Ang II-induced ET-1 transactivation. Endothelial-specific knockdown of Brg1/Brm or Ash2/Wdr5 ameliorated cardiac hypertrophy both in vitro and in vivo. More important, Brg1/Brm interacted with Ash2/Wdr5 on the ET-1 promoter to catalyze H3K4 methylation. The crosstalk between Brg11/Brm and Ash2/Wdr5 was mediated by myocardin-related transcription factor A (MRTF-A). In conclusion, our data have unveiled an epigenetic complex that links ET-1 transactivation in endothelial cells to Ang II-induced cardiac hypertrophy and fibrosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV36912
Anti-ASCL2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV36913
Anti-ASCL2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1305026
ANTI-ASCL2 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2104130
Anti-ASCL2 antibody produced in rabbit, affinity isolated antibody
SAB2106440
Anti-ASCL2 antibody produced in rabbit, affinity isolated antibody
SAB2100164
Anti-ASH2L (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100165
Anti-ASH2L (ab2) antibody produced in rabbit, affinity isolated antibody
AV34203
Anti-ASH2L antibody produced in rabbit, affinity isolated antibody
SAB4300685
Anti-ASH2L antibody produced in rabbit, affinity isolated antibody
HPA042289
Anti-ASH2L antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV37504
Anti-MKL1 antibody produced in rabbit, IgG fraction of antiserum
HPA030782
Anti-MKL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV34484
Anti-SMARCA2 antibody produced in rabbit, affinity isolated antibody
HPA029981
Anti-SMARCA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106546
Anti-WDR5 antibody produced in rabbit, affinity isolated antibody
SAB1404665
Monoclonal Anti-ASH2L antibody produced in mouse, clone 4G7, purified immunoglobulin, buffered aqueous solution
WH0006595M6
Monoclonal Anti-SMARCA2 antibody produced in mouse, clone 2D12, purified immunoglobulin, buffered aqueous solution
WH0011091M1
Monoclonal Anti-WDR5 antibody produced in mouse, clone 2C2, purified immunoglobulin, buffered aqueous solution
SAB1404900
Monoclonal Anti-WDR5 antibody produced in mouse, clone 2C2, ascites fluid
SAB1408817
Monoclonal Anti-WDR5 antibody produced in mouse, clone 1B1, purified immunoglobulin, buffered aqueous solution
PLA0028
Rabbit anti-ASH2 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0256
Rabbit anti-WDR5 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.