EMAIL THIS PAGE TO A FRIEND
The Journal of infectious diseases
A Potent Virus-Specific Antibody-Secreting Cell Response to Acute Enterovirus 71 Infection in Children.
PMID 25712974
Abstract
Enterovirus 71 (EV71) remains a leading pathogen for acute infectious diseases in children, especially in Asia. The cellular basis for establishing a virus-specific antibody response to acute EV71 infections is unclear in children. We studied the magnitude of virus-specific antibody-secreting B cells (ASCs) and its relationship with serological response, clinical parameters, and virological parameters among children with laboratory-confirmed EV71 infection. A potent EV71 genogroup B- and virus-specific ASC response was detected in the first week of illness among genotype B5 EV71-infected children. The cross-reactive EV71-specific ASC response to genogroup C viral antigens composed about 10% of the response. The EV71-specific ASC response in children aged ≥3 years produced immunoglobulin G predominantly, but immunoglobulin M was predominant in younger children. Proliferation marker was expressed by the majority of circulating ASCs in the acute phase of EV71 infection. Virus-specific ASC responses significantly correlated with throat viral load, fever duration, and serological genogroup-specific neutralization titer. The presence of a virus-specific ASC response serves an early cellular marker of an EV71-specific antibody response. Further detailed study of EV71-specific ASCs at the monoclonal level is crucial to delineate the specificity and function of antibody immunity in children.