EMAIL THIS PAGE TO A FRIEND
Oncotarget
ROS generation mediates the anti-cancer effects of WZ35 via activating JNK and ER stress apoptotic pathways in gastric cancer.
PMID 25714022
Abstract
Gastric cancer is one of the leading causes of cancer mortality in the world, and finding novel agents and strategies for the treatment of advanced gastric cancer is of urgent need. Curcumin is a well-known natural product with anti-cancer ability, but is limited by its poor chemical stability. In this study, an analog of curcumin with high chemical stability, WZ35, was designed and evaluated for its anti-cancer effects and underlying mechanisms against human gastric cancer. WZ35 showed much stronger anti-proliferative effects than curcumin, accompanied by dose-dependent induction of cell cycle arrest and apoptosis in gastric cancer cells. Mechanistically, our data showed that WZ35 induced reactive oxygen species (ROS) production, resulting in the activation of both JNK-mitochondrial and ER stress apoptotic pathways and eventually cell apoptosis in SGC-7901 cells. Blockage of ROS production totally reversed WZ35-induced JNK and ER stress activation as well as cancer cell apoptosis. In vivo, WZ35 showed a significant reduction in SGC-7901 xenograft tumor size in a dose-dependent manner. Taken together, this work provides a novel anticancer candidate for the treatment of gastric cancer, and importantly, reveals that increased ROS generation might be an effective strategy in human gastric cancer treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV38067
Anti-ATF4 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2501899
Anti-ATF4 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2500130
Anti-ATF4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2100168
Anti-ATF4 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2105294
Anti-ATF4, (C-terminal) antibody produced in rabbit, affinity isolated antibody
PRS3681
Anti-ATF6 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3683
Anti-ATF6 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1400432
Anti-ATF6 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2100170
Anti-ATF6 antibody produced in rabbit, affinity isolated antibody
HPA005935 Anti-ATF6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106254
Anti-ATF6 antibody produced in rabbit, affinity isolated antibody
AV30161
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
SAB2100360
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
SAB2100378
Anti-CDC2 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100379
Anti-CDC2 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2103891
Anti-CDC2 antibody produced in rabbit, affinity isolated antibody
SAB4500051
Anti-CDC2 antibody produced in rabbit, affinity isolated antibody
SAB4503707
Anti-CDC2 antibody produced in rabbit, affinity isolated antibody
HPA003387
Anti-CDK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
C8831
Anti-Cyclin B1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4503501
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
SAB4503502
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
SAB1405706
Anti-DDIT3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4500632
Anti-GADD153 antibody produced in rabbit, affinity isolated antibody
SAB3500381
Anti-XBP-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
GW21291 Anti-XBP1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22509 Anti-XBP1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV38552
Anti-XBP1 (AB2) antibody produced in rabbit, affinity isolated antibody
SAB1406598
Anti-XBP1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2102720
Anti-XBP1 antibody produced in rabbit, affinity isolated antibody