EMAIL THIS PAGE TO A FRIEND
Oncotarget
miR-188-5p inhibits tumour growth and metastasis in prostate cancer by repressing LAPTM4B expression.
PMID 25714029
Abstract
Elucidation of the molecular targets and pathways regulated by the tumour-suppressive miRNAs can shed light on the oncogenic and metastatic processes in prostate cancer (PCa). Using miRNA profiling analysis, we find that miR-188-5p was significantly down-regulated in metastatic PCa. Down-regulation of miR-188-5p is an independent prognostic factor for poor overall and biochemical recurrence-free survival. Restoration of miR-188-5p in PCa cells (PC-3 and LNCaP) significantly suppresses proliferation, migration and invasion in vitro and inhibits tumour growth and metastasis in vivo. We also find overexpression of miR-188-5p in PC-3 cells can significantly enhance the cells' chemosensitivity to adriamycin. LAPTM4B is subsequently identified as a direct target of miR-188-5p in PCa, and is found to be significantly over-expressed in PCa. Knockdown of LAPTM4B phenotypically copies miR-188-5p-induced phenotypes, whereas ectopic expression of LAPTM4B reverses the effects of miR-188-5p. We also find that restoration of miR-188-5p can inhibit the PI3K/AKT signaling pathway via the suppression of LAPTM4B. Taken together, this is the first report unveils that miR-188-5p acts as a tumour suppressor in PCa and may therefore serve as a useful therapeutic target for the development of new anticancer therapy.