EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Yeast Hmt1 catalyses asymmetric dimethylation of histone H3 arginine 2 inxa0vitro.
PMID 25715670
Abstract
Protein arginine methyltransferases (PRMTs) are a family of enzymes that can methylate protein arginine residues. PRMTs' substrates include histones and a variety of non-histone proteins. Previous studies have shown that yeast Hmt1 is a typexa0I PRMT and methylates histone H4 arginine 3 and several mRNA-binding proteins. Hmt1 forms dimers or oligomers, but how dimerization or oligomerization affects its activity remains largely unknown. We now report that Hmt1 can methylate histone H3 arginine 2 (H3R2) inxa0vitro. The dimerization but not hexamerization is essential for Hmt1's activity. Interestingly, the methyltransferase activity of Hmt1 on histone H3R2 requires reciprocal contributions from two Hmt1 molecules. Our results suggest an intermolecular trans-complementary mechanism by which Hmt1 dimer methylates its substrates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1822
Doxycycline monohydrate
C22H24N2O8 · H2O