EMAIL THIS PAGE TO A FRIEND
Circulation. Arrhythmia and electrophysiology
Synchronous systolic subcellular Ca2+-elevations underlie ventricular arrhythmia in drug-induced long QT type 2.
PMID 25722252
Abstract
Repolarization delay is a common clinical problem, which can promote ventricular arrhythmias. In myocytes, abnormal sarcoplasmic reticulum Ca(2+)-release is proposed as the mechanism that causes early afterdepolarizations, the cellular equivalent of ectopic-activity in drug-induced long-QT syndrome. A crucial missing link is how such a stochastic process can overcome the source-sink mismatch to depolarize sufficient ventricular tissue to initiate arrhythmias. Optical maps of action potentials and Ca(2+)-transients from Langendorff rabbit hearts were measured at low (150×150 μm(2)/pixel) and high (1.5×1.5 μm(2)/pixel) resolution before and during arrhythmias. Drug-induced long QT type 2, elicited with dofetilide inhibition of IKr (the rapid component of rectifying K+ current), produced spontaneous Ca(2+)-elevations during diastole and systole, before the onset of arrhythmias. Diastolic Ca(2+-)waves appeared randomly, propagated within individual myocytes, were out-of-phase with adjacent myocytes, and often died-out. Systolic secondary Ca(2+-)elevations were synchronous within individual myocytes, appeared 188±30 ms after the action potential-upstroke, occurred during high cytosolic Ca(2+) (40%-60% of peak-Ca(2+)-transients), appeared first in small islands (0.5×0.5 mm(2)) that enlarged and spread throughout the epicardium. Synchronous systolic Ca(2+-)elevations preceded voltage-depolarizations (9.2±5 ms; n=5) and produced pronounced Spatial Heterogeneities of Ca(2+)-transient-durations and action potential-durations. Early afterdepolarizations originating from sites with the steepest gradients of membrane-potential propagated and initiated arrhythmias. Interestingly, more complex subcellular Ca(2+)-dynamics (multiple chaotic Ca(2+)-waves) occurred during arrhythmias. K201, a ryanodine receptor stabilizer, eliminated Ca(2+)-elevations and arrhythmias. The results indicate that systolic and diastolic Ca(2+)-elevations emanate from sarcoplasmic reticulum Ca(2+)-release and systolic Ca(2+)-elevations are synchronous because of high cytosolic and luminal-sarcoplasmic reticulum Ca(2+), which overcomes source-sink mismatch to trigger arrhythmias in intact hearts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0016
Dofetilide, ≥98% (HPLC)
C19H27N3O5S2