PloS one
Raltegravir non-inferior to nucleoside based regimens in second-line therapy with lopinavir/ritonavir over 96 weeks: a randomised open label study for the treatment of HIV-1 infection.
PMID 25723472
Abstract
To determine the durability over 96 weeks of safety and efficacy of lopinavir/ritonavir (LPV/r) and raltegravir (RAL) which was demonstrated to have non-inferior efficacy relative to a regimen of LPV/r with nucleoside/nucleotide reverse transcriptase inhibitors (N(t)RTIs) (Control) in primary analysis at 48 weeks. Open label, centrally randomised trial. Recruitment was from 37 primary and secondary care sites from Africa, Asia, Australia, Europe and Latin America. 541 HIV-1 infected adults virologically failing first-line non-NRTI + 2N(t)RTI, with no previous exposure to protease inhibitors or integrase strand transfer inhibitors were analysed, 425 completed 96 weeks follow up on randomised therapy. Randomisation was 1:1 to Control or RAL. Differences between the proportion of participants with plasma HIV-1 RNA (VL) <200 copies/mL by intention to treat were compared with a non-inferiority margin of -12%. Differences in biochemical, haematological and metabolic changes were assessed using T-tests. VL <200 copies/mL at 96 weeks was: RAL 80.4%, Control 76.0% (difference: 4.4 [95%CI -2.6, 11.3]) and met non-inferiority criteria. The RAL arm had a significantly higher mean change (difference Control-RAL; 95%CI) in haemoglobin (-2.9; -5.7, -1.1), total lymphocytes (-0.2; -0.3, -0.0), total cholesterol (-0.5; -0.8, -0.3), HDL cholesterol (-0.1; -0.1, -0.0) and LDL cholesterol (-0.3; -0.5, -0.2). At 96 weeks, both RAL and Control maintained efficacy greater than 75% and continued to demonstrate similar safety profiles. These results support the use of a combination LPV/r and RAL regimen as an option following failure of 1st line NNRTI + 2N(t)RTIs. ClinicalTrials.gov NCT00931463.