EMAIL THIS PAGE TO A FRIEND
Cancer research
RIP1 Kinase Is an Oncogenic Driver in Melanoma.
PMID 25724678
Abstract
Although many studies have uncovered an important role for the receptor-binding protein kinase RIP1 in controlling cell death signaling, its possible contributions to cancer pathogenesis have been little explored. Here, we report that RIP1 functions as an oncogenic driver in human melanoma. Although RIP1 was commonly upregulated in melanoma, RIP1 silencing inhibited melanoma cell proliferation in vitro and retarded the growth of melanoma xenografts in vivo. Conversely, while inducing apoptosis in a small proportion of melanoma cells, RIP1 overexpression enhanced proliferation in the remaining cells. Mechanistic investigations revealed that the proliferative effects of RIP1 overexpression were mediated by NF-κB activation. Strikingly, ectopic expression of RIP1 enhanced the proliferation of primary melanocytes, triggering their anchorage-independent cell growth in an NF-κB-dependent manner. We identified DNA copy-number gain and constitutive ubiquitination by a TNFα autocrine loop mechanism as two mechanisms of RIP1 upregulation in human melanomas. Collectively, our findings define RIP1 as an oncogenic driver in melanoma, with potential implications for targeting its NF-κB-dependent activation mechanism as a novel approach to treat this disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA005513
Anti-BIRC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401012
Anti-BIRC2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501559
Anti-BIRC3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501560
Anti-BIRC3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1409216
Anti-BIRC3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002317
Anti-BIRC3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401013
Anti-BIRC3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA046651
Anti-RALBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410141
Anti-RALBP1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2105823
Anti-RALBP1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
GW21168 Anti-RIPK1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA015257
Anti-RIPK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV02038
Anti-RIPK1 antibody produced in rabbit, affinity isolated antibody
SAB3500420
Anti-RIPK1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1401396
Anti-UQCRFS1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401397
Anti-UQCRFS1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA041863
Anti-UQCRFS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105078
Anti-UQCRFS1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4100427
Monoclonal Anti-BIRC3 antibody produced in mouse, clone BIRC3-A359, culture supernatant
WH0010928M2
Monoclonal Anti-RALBP1 antibody produced in mouse, clone 2A1, purified immunoglobulin, buffered aqueous solution
SAB1404880 Monoclonal Anti-RALBP1 antibody produced in mouse, clone 1C2-A10, purified immunoglobulin, buffered aqueous solution
SAB4100176
Monoclonal Anti-RALBP1 antibody produced in mouse, clone RALBP1-A192, culture supernatant
SRP3177
TNF-α human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
T0157
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in yeast, buffered aqueous solution, suitable for cell culture
T6674
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
H8916
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture