EMAIL THIS PAGE TO A FRIEND
Diabetes research and clinical practice
Marker of lipid peroxidation related to diabetic nephropathy in Indonesian type 2 diabetes mellitus patients.
PMID 25726335
Abstract
Even though diabetes patients exhibit an increased oxidative stress, its correlation with diabetic nephropathy is not fully understood. The purpose of this study was to determine whether lipid peroxidation marker correlates well with eGFR and UACR in type 2 diabetes mellitus patients. We collected urine and serum samples of Indonesian type 2 diabetes mellitus outpatients with normo- and microalbuminuria at a Local Government Clinic (from ages: 39-74 years). Urinary 8-iso-PGF2α was measured by ELISA, the serum malondialdehyde by TBARS assay, and urinary albumin by BCG albumin assay. eGFR was calculated using the corrected-Cockcroft-Gault (CG), MDRD, and CKD-EPI equation. Other necessary data were obtained through questionnaires. The results showed that the increasing level of malondialdehyde was mildly correlated with the decline in eGFR (MDRD). In contrary, there was a significant positive correlation between 8-iso-PGF2α concentration and eGFR based on the corrected-CG, MDRD study, and CKD-EPI equation (r=0.457, p<0.001; r=0.424, p<0.001; r=0.443, p<0.001). This relationship still persisted in the normoalbuminuric subjects (n=43) (r=0.491, p=0.001; r=0.461, p=0.002; r=0.455, p=0.002). The multivariate analysis showed that 8-iso-PGF2α together with fasting plasma glucose was the most predictive factor for the high 2-quantile eGFR (adjusted OR 1.001, (95% CI, 1.000-1.001)). However, there was no significant correlation between UACR with malondialdehyde (r=0.268, p=0.050) and 8-iso-PGF2α(r=-0.030, p=0.808). UACR itself was inversely correlated with eGFR based on the corrected-CG, the MDRD, and CKD-EPI (r=-0.232, p<0.05; r=-0.228, p<0.05; r=-0.232, p<0.05). Increased 8-iso-PGF2α and malondialdehyde in type 2 diabetes mellitus patients may play a role in the pathophysiologic significance of diabetic nephropathy, even while considering the effect of potential confounders.