EMAIL THIS PAGE TO A FRIEND
Journal of insect physiology
Juvenile hormone (JH) esterase activity but not JH epoxide hydrolase activity is downregulated in larval Adoxophyes honmai following nucleopolyhedroviruses infection.
PMID 25727179
Abstract
Juvenile hormones (JHs) and ecdysteroids are critical insect developmental hormones. JH esterase (JHE) and JH epoxide hydrolase (JHEH) are JH-selective enzymes that metabolize JH and thus regulate the titer of JH. Baculoviruses are known to alter host endocrine regulation. The nucleopolyhedroviruses, AdhoNPV and AdorNPV, are known to have slow and fast killing activity against Adoxophyes honmai (Lepidoptera: Tortricidae), respectively. Here we found that when penultimate (4th) instar A. honmai are inoculated with AdhoNPV or AdorNPV, the mean survival time is 9.7 and 8.2 days, respectively. The larvae molted once but did not pupate. The AdhoNPV- or AdorNPV-infected larvae did not show a dramatic increase in JHE activity as was found in mock-infected larvae, instead they showed a marked decrease in JHE activity. In contrast, both viral infections had no effect on JHEH activity. In order to further characterize the JHE activity, the JHE-coding sequence of A. honmai (ahjhe) was cloned and confirmed to encode a biologically active JHE. Quantitative real-time PCR analysis of ahjhe expression in 4th and 5th instar A. honmai revealed that AdhoNPV and AdorNPV are able to reduce ahjhe expression levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0879
Diisopropylfluorophosphate
C6H14FO3P
J2000
Juvenile hormone III, ≥65%
C16H26O3