EMAIL THIS PAGE TO A FRIEND
Journal of animal breeding and genetics = Zeitschrift fur Tierzuchtung und Zuchtungsbiologie
A retrospective analysis of allele frequency changes of major genes during 20xa0years of selection in the Italian Large White pig breed.
PMID 25727360
Abstract
In this study, we investigated whether a selection programme based on boar genetic evaluation obtained with a classical BLUP animal model can change allele frequencies in a pig population. All Italian Large White boars born from 1992 to 2012 with estimated breeding value reliability >0.85 (nxa0=xa0200) were selected among all boars of this breed. Boars were genotyped with markers in major genes (IGF2 intron3-g.3072G>A, MC4R p.D298N, VRTN PRE1 insertion, PRKAG3 p.I199V and FTO g.276T>G). Genotyping data were analysed grouping boars in eight classes according to their year of birth. To evaluate the influence of time on allele frequencies of the genotyped markers, multinomial logistic regression models were computed. Four of five polymorphic sites (IGF2, MC4R, VRTN and FTO) showed significant (pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T19607
1,2,4,5-Tetramethylbenzene, 98%
C10H14