EMAIL THIS PAGE TO A FRIEND
Clinical immunology (Orlando, Fla.)
Ex vivo peptide-MHC II tetramer analysis reveals distinct end-differentiation patterns of human pertussis-specific CD4(+) T cells following clinical infection.
PMID 25728491
Abstract
Pertussis is occurring in highly vaccinated populations, suggesting insufficient protective memory CD4(+) T cells to Bordetella (B.) pertussis. P.69 Pertactin (P.69 Prn) is an important virulence factor of B. pertussis, and P.69 Prn7-24 is an immunodominant CD4(+) T cell epitope in mice and broadly recognized in humans. P.69 Prn7-24 peptide-MHC II tetramers (DRB4*0101/IVKT) were designed to ex vivo interrogate the presence and differentiation state of P.69 Prn7-24 specific CD4(+) T cells in six symptomatic pertussis cases. Cases with relatively more CD45RA(-)CCR7(+) central memory CD4(+)DRB4*0101/IVKT(+) T cells secreted Th1 cytokines, while cases with more CD45RA(-)CCR7(-) effector memory CD4(+)DRB4*0101/IVKT(+) T cells secreted both Th1 and Th2 cytokines upon peptide stimulation. CD45RA(+)CCR7(-) terminal differentiation pattern was associated with low or non-functionality based on cytokine secretion. This study provides proof of principle for further peptide-MHC II tetramer guided approaches in the elucidation of limited immunological memory to B. pertussis and the resurgence of pertussis.
Related Materials