EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Renewable-juglone-based high-performance sodium-ion batteries.
PMID 25728939
Abstract
A renewable-biomolecule-based electrode is developed through a facile synchronous reduction and self-assembly process, without any binder or additional conductive agent. The hybridized electrodes can be fabricated with arbitrary size and shape and exhibit superior capacity and cycle performance. The renewable-biomaterial-based high-performance electrodes will hold a place in future energy-storage devices.