EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Identification and characterization of the aroma-impact components of Thai fish sauce.
PMID 25730550
Abstract
Comprehensive analysis of the potent odorants in Thai premium fish sauce samples was accomplished by use of complementary volatile isolation methods combined with gas chromatography-olfactometry (GC-O) and GC-mass spectrometry. Odorants of intermediate and low volatility were determined by direct solvent extraction/solvent-assisted flavor evaporation (DSE-SAFE) and aroma extract dilution analysis (AEDA). Meanwhile, static headspace dilution analysis (SHDA) and headspace solid-phase microextraction (H-SPME) were used to determine the highly volatile odorants. Results of AEDA indicated the importance (log3FD factor≥6) of five acidic odorants (butanoic acid, 3-methylbutanoic acid, 4-hydroxy-2,5-dimethyl-3(2H)-furanone, 4-hydroxy-2-ethyl-5-methyl-3(2H)-furanone, and 2-phenylacetic acid) and four neutral/basic odorants (3-methylbutanal, (Z)-1,5-octadien-3-one, phenylacetaldehyde, and o-aminoacetophone). Results of SHDA indicated the predominant (log3FD factors≥5) headspace odorants were methanethiol, 2-methylpropanal, 2-methylbutanal, 3-methylbutanal, dimethyl trisulfide, 3-(methylthio)propanal, and butanoic acid. Concentrations for 21 odorants were determined by stable isotope dilution analysis (SIDA), and their odor-activity values (OAVs) were calculated. Among these, methanethiol, 2-methylpropanal, 3-methylbutanal, dimethyl trisulfide, 3-(methylthio)propanal, and butanoic acid had the highest OAVs (>500). Results of aroma recombination and omission studies revealed the importance of acids, aldehydes, and sulfur-containing compounds to the overall aroma of the Thai fish sauce.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
40703
4-Hydroxy-2,5-dimethyl-3(2H)-furanone, ≥99.0% (GC)
C6H8O3