EMAIL THIS PAGE TO A FRIEND
Nature neuroscience
Fast clonal expansion and limited neural stem cell self-renewal in the adult subependymal zone.
PMID 25730673
Abstract
We analyzed the progeny of individual neural stem cells (NSCs) of the mouse adult subependymal zone (SEZ) in vivo and found a markedly fast lineage amplification, as well as limited NSC self-renewal and exhaustion in a few weeks. We further unraveled the mechanisms of neuronal subtype generation, finding that a higher proportion of NSCs were dedicated to generate deep granule cells in the olfactory bulb and that larger clones were produced by these NSCs.