EMAIL THIS PAGE TO A FRIEND
Journal of medicinal food
Acute effects of black currant consumption on salivary flow rate and secretion rate of salivary immunoglobulin a in healthy smokers.
PMID 25734687
Abstract
The role of saliva in maintaining oral health and homeostasis is based on its physicochemical properties and biological activities of its components, including salivary immunoglobulin A (IgA). Both salivary rates and immunological status of saliva are found to be compromised in smokers. The aim of this study was to investigate the acute time-dependent effect of smoking and black currant consumption on the salivary flow rate (SFR) and salivary IgA secretion rate (sIgA SR) in healthy smokers. SFR, sIgA levels in saliva, and sIgA SRs were determined in healthy smokers (n=8) at eight times of assessment within three consecutive interventions: at the baseline; 5, 30, and 60 min after smoking; 5, 30, and 60 min after black currant consumption (100 g), followed by smoking; and 5 min after black currant consumption. Smoking induced a significant delayed effect on SFR measured 60 min after smoking (P=.03), while black currant consumption preceding smoking prevented that effect. Salivary IgA concentrations and sIgA flow rates were not acutely influenced by smoking. Black currant consumption preceding smoking induced a significant decrease in sIgA concentrations 5 min after the intervention compared with the baseline (P=.046), with a further increasing trend, statistically significant, 60 min after the intervention (P=.025). Although smoking cessation is the most important strategy in the prevention of chronic diseases, the obtained results suggest that the influence of black currant consumption on negative effects of tobacco smoke on salivary flow and immunological status of saliva could partly reduce the smoking-associated risk on oral health.