EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
The Characterization of Varicella Zoster Virus-Specific T Cells in Skin and Blood during Aging.
PMID 25734814
Abstract
Reactivation of the varicella zoster virus (VZV) increases during aging. Although the effects of VZV reactivation are observed in the skin (shingles), the number and functional capacity of cutaneous VZV-specific T cells have not been investigated. The numbers of circulating IFN-γ-secreting VZV-specific CD4(+) T cells are significantly decreased in old subjects. However, other measures of VZV-specific CD4(+) T cells, including proliferative capacity to VZV antigen stimulation and identification of VZV-specific CD4(+) T cells with an major histocompatibility complex class II tetramer (epitope of IE-63 protein), were similar in both age groups. The majority of T cells in the skin of both age groups expressed CD69, a characteristic of skin-resident T cells. VZV-specific CD4(+) T cells were significantly increased in the skin compared with the blood in young and old subjects, and their function was similar in both age groups. In contrast, the number of Foxp3(+) regulatory T cells and expression of the inhibitory receptor programmed cell death -1 PD-1 on CD4(+) T cells were significantly increased in the skin of older humans. Therefore, VZV-specific CD4(+) T cells in the skin of older individuals are functionally competent. However, their activity may be restricted by multiple inhibitory influences in situ.