EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section C, Structural chemistry
Structure determination and modeling of monoclinic trioctylphosphine oxide.
PMID 25734856
Abstract
Trioctylphosphine oxide (TOPO), C(24)H(51)OP, was recrystallized from ambient evaporation in acetone. TOPO single crystals form with a monoclinic P2(1)/c structure. Fourier transform IR (FT-IR) spectroscopy captures the characteristic stretching modes from the seven methylene groups, the phosphoryl P=O bond, and the phosphoryl-carbon bond.