EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Targeting of the deubiquitinase USP9X attenuates B-cell acute lymphoblastic leukemia cell survival and overcomes glucocorticoid resistance.
PMID 25735983
Abstract
Although previous studies attributed a pro-survival role to USP9X in human cancer, how USP9X affects B-cell acute lymphoblastic leukemia (B-ALL) remains unclear. Here, we found that USP9X is overexpressed in B-ALL cell lines and human patients. We investigated the role of USP9X in B-ALL and found that USP9X knockdown significantly reduced leukemic cell growth and increased spontaneous apoptosis, thereby improving survival in immunodeficient mice. These effects are partially mediated by the intrinsic apoptotic pathway, as we found that USP9X-knockdown leukemic cells displayed MCL1 down-regulation, with decreased BCL-2/BCL-XL levels and increased BAX levels. In addition, we demonstrated that USP9X inhibition negatively regulates mTORC1 activity toward its substrate S6K1. Clinically, USP9X inhibition sensitized glucocorticoid-resistant ALL cells to prednisolone; this observation reveals a potential avenue for improving the treatment of drug-resistant relapses. Collectively, our findings suggest that the combination of USP9X targeting and glucocorticoids treatment has attractive utility in B-ALL. This approach represents a potential strategy for promising combination therapies for lymphoid malignancies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2107379
Anti-USP9X antibody produced in rabbit, affinity isolated antibody