EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Covalent and stable CuAAC modification of silicon surfaces for control of cell adhesion.
PMID 25737226
Abstract
Stable primary functionalization of metal surfaces plays a significant role in reliable secondary attachment of complex functional molecules used for the interfacing of metal objects and nanomaterials with biological systems. In principle, this can be achieved through chemical reactions either in the vapor or liquid phase. In this work, we compared these two methods for oxidized silicon surfaces and thoroughly characterized the functionalization steps by tagging and fluorescence imaging. We demonstrate that the vapor-phase functionalization only provided transient surface modification that was lost on extensive washing. For stable surface modification, a liquid-phase method was developed. In this method, silicon wafers were decorated with azides, either by silanization with (3-azidopropyl)triethoxysilane or by conversion of the amine groups of an aminopropylated surface by means of the azido-transfer reaction. Subsequently, D-amino acid adhesion peptides could be immobilized on the surface by use of Cu(I)-catalyzed click chemistry. This enabled the study of cell adhesion to the metal surface. In contrast to unmodified surfaces, the peptide-modified surfaces were able to maintain cell adhesion during significant flow velocities in a microflow reactor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8027
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
CuO4S · 5H2O
C3036
Copper(II) sulfate pentahydrate, plant cell culture tested, ≥98%
CuO4S · 5H2O
203165
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
CuO4S · 5H2O
469130
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
CuO4S · 5H2O
209198
Copper(II) sulfate pentahydrate, ACS reagent, ≥98.0%
CuO4S · 5H2O
C7631
Copper(II) sulfate pentahydrate, ReagentPlus®, ≥98.0%
CuO4S · 5H2O
31293
Copper(II) sulfate pentahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99-102%
CuO4S · 5H2O
61245
Copper(II) sulfate pentahydrate, purum p.a., crystallized, ≥99.0% (RT)
CuO4S · 5H2O
12849
Copper(II) sulfate pentahydrate, puriss., suitable for, meets analytical specification of Ph. Eur., BP, USP,FCC
CuO4S · 5H2O
RES10395-B7
Cupric sulfate pentahydrate, PharmaGrade, EP, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
CuO4S · 5H2O
294594
Mercury, electronic grade, 99.9999% trace metals basis
Hg
03440
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
C8H19N
03439
N-Ethyldiisopropylamine solution, for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
C8H19N
91737
Trifluoromethanesulfonic anhydride, purum, ≥98.0% (T)
C2F6O5S2
176176
Trifluoromethanesulfonic anhydride, ≥99%
C2F6O5S2
704083
Trifluoromethanesulfonic anhydride solution, 1 M in methylene chloride
C2F6O5S2