EMAIL THIS PAGE TO A FRIEND
BioMed research international
Acoustic-frequency vibratory stimulation regulates the balance between osteogenesis and adipogenesis of human bone marrow-derived mesenchymal stem cells.
PMID 25738155
Abstract
Osteoporosis can be associated with the disordered balance between osteogenesis and adipogenesis of bone marrow-derived mesenchymal stem cells (BM-MSCs). Although low-frequency mechanical vibration has been demonstrated to promote osteogenesis, little is known about the influence of acoustic-frequency vibratory stimulation (AFVS). BM-MSCs were subjected to AFVS at frequencies of 0, 30, 400, and 800 Hz and induced toward osteogenic or adipogenic-specific lineage. Extracellular matrix mineralization was determined by Alizarin Red S staining and lipid accumulation was assessed by Oil Red O staining. Transcript levels of osteogenic and adipogenic marker genes were evaluated by real-time reverse transcription-polymerase chain reaction. Cell proliferation of BM-MSCs was promoted following exposure to AFVS at 800 Hz. Vibration at 800 Hz induced the highest level of calcium deposition and significantly increased mRNA expression of COL1A1, ALP, RUNX2, and SPP1. The 800 Hz group downregulated lipid accumulation and levels of adipogenic genes, including FABP4, CEBPA, PPARG, and LEP, while vibration at 30 Hz supported adipogenesis. BM-MSCs showed a frequency-dependent response to acoustic vibration. AFVS at 800 Hz was the most favorable for osteogenic differentiation and simultaneously suppressed adipogenesis. Thus, acoustic vibration could potentially become a novel means to prevent and treat osteoporosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1300000
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
C6H8O6
1043003
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
C6H8O6
PHR1008
Ascorbic Acid
C6H8O6
03378
Chloride atomic spectroscopy standard concentrate 10.00 g Cl-, 10.00 g/L, for 1 l standard solution, analytical standard
HCl
12156
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
H2O
D1756
Dexamethasone, ≥98% (HPLC), powder
C22H29FO5
D9184
Dexamethasone, meets USP testing specifications
C22H29FO5
D0700000
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
1176007
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
C22H29FO5
46165
Dexamethasone, VETRANAL, analytical standard
C22H29FO5
31375
Dexamethasone, tested according to Ph.Eur.
C22H29FO5
PHR1526
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
C22H29FO5
Y0001593
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
Y0001177
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
2107
E-Toxate Water, endotoxin, free
H2O
47673
Formaldehyde solution, tested according to Ph.Eur.
CH2O
84426
Hydrochloric acid, purum p.a., fuming 37%, ≥37% (T)
HCl
17933
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization
HCl
17934
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization
HCl
17935
Hydrogen chloride – methanol solution, ~1.25 M HCl, for GC derivatization
HCl
I7378
Indomethacin, ≥99% (TLC)
C19H16ClNO4
I8280
Indomethacin, meets USP testing specifications
C19H16ClNO4
1341001
Indomethacin, United States Pharmacopeia (USP) Reference Standard
C19H16ClNO4
I0200000
Indomethacin, European Pharmacopoeia (EP) Reference Standard
C19H16ClNO4
PHR1247
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
C19H16ClNO4
57803
L-Ascorbic acid, certified reference material, TraceCERT®
C6H8O6
95212
L-Ascorbic acid, tested according to Ph.Eur.
C6H8O6
47863
L-Ascorbic acid, analytical standard
C6H8O6
DENWAT
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
H2O
DENWAT3
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
H2O