EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Conditioned medium from umbilical cord mesenchymal stem cells induces migration and angiogenesis.
PMID 25739039
Abstract
Umbilical cord mesenchymal stem cells (UC-MSCs) have been suggested as a candidate for various clinical applications, however, major limitations include the lack of organ-specific accumulation and low survival rates of transplanted cells. In the present study, it was hypothesized that the paracrine effects of UC‑MSCs may enhance stem cell-based tissue repair and regeneration by promoting the specific homing of stem/progenitor cells and the overall ability to drive them to the damaged area. UC-MSCs-derived conditioned medium (UC-CM) was analyzed using liquid chip and ELISA techniques. In vitro tube formation assays of human umbilical vein endothelial cells (HUVECs) and UC-MSCs were then performed to assess the angiogenic properties of UC-CM. Subsequently, UC-MSCs, HUVECs and fibroblasts were labeled with PKH26 for an in vivo cell migration assay. The expression levels of C-X-C chemokine receptor 4 (CXCR4), C-C chemokine receptor 2 (CCR2) and c-met were determined in the UC-MSCs, HUVECs and fibroblasts using reverse transcription-quantitative polymerase chain reaction and flow cytometry. UC-CM was incubated with or without antibodies, and the contribution of stromal cell-derived factor 1 (SDF-1), monocyte chemotactic protein 1 (MCP-1) and hepatocyte growth factor (HGF) on the migration of cells was investigated in vitro. The results demonstrated that UC-MSCs secreted different cytokines and chemokines, including increased quantities of SDF-1, MCP-1 and HGF, in addition to the angiogenic factors, vascular cell adhesion protein-1, interleukin-8, insulin-like growth factor-1 and vascular endothelial growth factor. The total lengths of the tubes were significantly increased in the UC-MSCs and HUVECs incubated in UC-CM compared with those incubated in Dulbecco's modified Eagle's medium. In vivo cell migration assays demonstrated that UC-CM was a chemotactic stimulus for the UC-MSCs and HUVECs. In vitro Matrigel migration and scratch healing assays demonstrated that UC-CM increased the migration of CXCR4-positive or/and CCR2-positive cells in a dose-dependent manner. In addition, different molecules were screened under antibody-based blocking migration conditions. The data revealed that the SDF-1/CXCR4 and MCP-1/CCR2 axes were involved in the chemoattractive activity of UC-CM and suggested that the effective paracrine factor of UC-CM is a large complex rather than a single factor. The results of the present study supported the hypothesis that UC-MSCs release soluble factors, which may extend the therapeutic applicability of stem cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1406413
Anti-CCL2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA019163
Anti-CCL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103318
Anti-CCL2 antibody produced in rabbit, affinity isolated antibody
SAB1411231
Anti-CCL2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
H7157 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H0652 Anti-Hepatocyte Growth Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
H0540 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H9786 Anti-Hepatocyte Growth Factor Receptor (c-Met) antibody produced in goat, affinity isolated antibody, lyophilized powder
H9911 Anti-Hepatocyte Growth Factor Receptor antibody produced in goat, lyophilized powder, affinity isolated antibody
SAB2101033
Anti-HGF (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101034
Anti-HGF (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2105571
Anti-HGF antibody produced in rabbit, affinity isolated antibody
HPA040360
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA044088
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410624
Anti-HGF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300599
Anti-MET (Ab-1003) antibody produced in rabbit, affinity isolated antibody
SAB4300479
Anti-MET (Ab-1234) antibody produced in rabbit, affinity isolated antibody
SAB4300487
Anti-MET (Ab-1349) antibody produced in rabbit, affinity isolated antibody
SAB1406117
Anti-MET antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501870
Anti-MET antibody produced in rabbit, affinity isolated antibody
SAB4501868
Anti-MET, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501869
Anti-MET, C-Terminal antibody produced in rabbit, affinity isolated antibody
15307
Chloral hydrate, meets analytical specification of Ph. Eur., BP, USP, 99.5-101%
C2H3Cl3O2
RAB0026
Human BDNF ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0212
Human HGF ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0213
Human HGF ELISA Kit, for cell and tissue lysates
RAB0676 Human MET / Hepatocyte Growth Factor Receptor ELISA Kit
RAB0123
Human SDF 1 α ELISA Kit, for plasma, cell culture supernatant and urine
RAB0124 Human SDF-1 β ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0505
Human VCAM-1 ELISA Kit, for serum, plasma, cell culture supernatant and urine