EMAIL THIS PAGE TO A FRIEND
Gene therapy
Tolerance of activated pathogenic CD4+ T cells by transcriptional targeting of dendritic cells.
PMID 25739989
Abstract
We have recently shown that targeted expression of myelin oligodendrocyte glycoprotein (MOG) to dendritic cells with self-inactivating-lentivirus vectors induces antigen-specific tolerance in naive antigen-specific CD4+ T cells and protects mice from experimental autoimmune encephalomyelitis (EAE). In the present study, we demonstrate that this approach also induces tolerance of activated antigen-specific CD4+ T cells and completely protects mice from passive EAE induction. Tolerance induction did not correlate with the depletion of the preactivated antigen-specific CD4+ T cells. However, upon isolation and in vitro re-stimulation at day 6 after adoptive transfer the MOG-specific CD4+ T cells from the non-tolerized mice produced large amounts of inflammatory cytokines, whereas those from tolerized mice did not. This unresponsiveness correlated with the upregulation of regulatory molecules associated with anergy and regulatory T cells (Tregs). The in vivo depletion of Tregs resulted in EAE susceptibility of the tolerized animals, suggesting that these cells have indeed a role in tolerance induction/maintenance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1300799
Anti-α-Synuclein (N-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
S3062
Anti-α-Synuclein antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
S5566
Anti-α-Synuclein antibody, Mouse monoclonal, clone Syn211, purified from hybridoma cell culture
SAB2500176
Anti-BTLA antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1412118
ANTI-BTLA antibody produced in mouse, clone 1B7, purified immunoglobulin, buffered aqueous solution
SAB1412119
ANTI-BTLA antibody produced in mouse, clone 2G8, purified immunoglobulin, buffered aqueous solution
HPA047211 Anti-BTLA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PRS4065
Anti-PD-1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4067
Anti-PD-1 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900280
Anti-PDCD1 (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900373
Anti-PDCD1 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2500763
Anti-PDCD1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1400196
Anti-PDCD1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA035981
Anti-PDCD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406423
Anti-SELL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4300502
Anti-SNCA (Ab-125) antibody produced in rabbit, affinity isolated antibody
SAB4300445
Anti-SNCA (Ab-129) antibody produced in rabbit, affinity isolated antibody
SAB4300528
Anti-SNCA (Ab-133) antibody produced in rabbit, affinity isolated antibody
SAB2502111
Anti-SNCA antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502112
Anti-SNCA antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2102253
Anti-SNCA antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA005459
Anti-SNCA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1105828
Anti-SPATA2 (341-354) antibody produced in rabbit, IgG fraction of antiserum
SAB1410026
Anti-SPATA2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA048581
Anti-SPATA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107365
Anti-SPATA2 antibody produced in rabbit, affinity isolated antibody
SAB1411697
Anti-SPATA2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SRP3242 IL-2 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
I0523 Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
I4161 Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, carrier free