EMAIL THIS PAGE TO A FRIEND
Glia
A novel role of microglial NADPH oxidase in mediating extra-synaptic function of norepinephrine in regulating brain immune homeostasis.
PMID 25740080
Abstract
Although the peripheral anti-inflammatory effect of norepinephrine (NE) is well documented, the mechanism by which this neurotransmitter functions as an anti-inflammatory/neuroprotective agent in the central nervous system (CNS) is unclear. This article aimed to determine the anti-inflammatory/neuroprotective effects and underlying mechanisms of NE in inflammation-based dopaminergic neurotoxicity models. In mice, NE-depleting toxin N-(2-chloroethyl)-N-ethyl-2-bromobenzylamine (DSP-4) was injected at 6 months of lipopolysaccharide (LPS)-induced neuroinflammation. It was found that NE depletion enhanced LPS-induced dopaminergic neuron loss in the substantia nigra. This piece of in vivo data prompted us to conduct a series of studies in an effort to elucidate the mechanism as to how NE affects dopamine neuron survival by using primary midbrain neuron/glia cultures. Results showed that submicromolar concentrations of NE dose-dependently protected dopaminergic neurons from LPS-induced neurotoxicity by inhibiting microglia activation and subsequent release of pro-inflammatory factors. However, NE-elicited neuroprotection was not totally abolished in cultures from β2-adrenergic receptor (β2-AR)-deficient mice, suggesting that novel pathways other than β2-AR are involved. To this end, It was found that submicromolar NE dose-dependently inhibited NADPH oxidase (NOX2)-generated superoxide, which contributes to the anti-inflammatory and neuroprotective effects of NE. This novel mechanism was indeed adrenergic receptors independent since both (+) and (-) optic isomers of NE displayed the same potency. We further demonstrated that NE inhibited LPS-induced NOX2 activation by blocking the translocation of its cytosolic subunit to plasma membranes. In summary, we revealed a potential physiological role of NE in maintaining brain immune homeostasis and protecting neurons via a novel mechanism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA019773
Anti-AKAP13 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408667
Anti-AKAP13 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4500229
Anti-AKAP13 antibody produced in rabbit, affinity isolated antibody
SAB2700772
Anti-DDX39B antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700773
Anti-DDX39B antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV50512
Anti-ING1 antibody produced in rabbit, IgG fraction of antiserum
SAB5200085 Anti-ING1 antibody produced in rabbit, 250 μg/mL, affinity isolated antibody
SAB1410716
Anti-ING1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA050628
Anti-NSFL1C antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA047108
Anti-NSFL1C antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411092
Anti-NSFL1C antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV32379
Anti-RUVBL2 (AB1) antibody produced in rabbit, affinity isolated antibody
AV32378
Anti-RUVBL2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
L3012 Lipopolysaccharides from Escherichia coli O111:B4, purified by gel-filtration chromatography