EMAIL THIS PAGE TO A FRIEND
Bone
Cbl-b and c-Cbl negatively regulate osteoblast differentiation by enhancing ubiquitination and degradation of Osterix.
PMID 25744063
Abstract
E3 ubiquitin ligase Cbl-b and c-Cbl play important roles in bone formation and maintenance. Cbl-b and c-Cbl regulate the activity of various receptor tyrosine kinases and intracellular protein tyrosine kinases mainly by regulating the degradation of target proteins. However, the precise mechanisms of how Cbl-b and c-Cbl regulate osteoblast differentiation are not well known. In this study, we investigated potential targets of Cbl-b and c-Cbl. We found that Cbl-b and c-Cbl inhibit BMP2-induced osteoblast differentiation in mesenchymal cells. Among various osteogenic transcription factors, we identified that Cbl-b and c-Cbl suppress the protein stability and transcriptional activity of Osterix. Our results suggest that Cbl-b and c-Cbl inhibit the function of Osterix by enhancing the ubiquitin-proteasome-mediated degradation of Osterix. Taken together, we propose novel regulatory roles of Cbl-b and c-Cbl during osteoblast differentiation in which Cbl-b and c-Cbl regulate the degradation of Osterix through the ubiquitin-proteasome pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5533
Alizarin Red S, certified by the Biological Stain Commission
C14H7NaO7S