EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Dextran-based hydrogel formed by thiol-Michael addition reaction for 3D cell encapsulation.
PMID 25744162
Abstract
Cell encapsulation in three-dimensional (3D) hydrogels can mimic native cell microenvironment and plays a major role in cell-based transplantation therapies. In this contribution, a novel in situ-forming hydrogel, Dex-l-DTT hydrogel ("l" means "linked-by"), by cross-linking glycidyl methacrylate derivatized dextran (Dex-GMA) and dithiothreitol (DTT) under physiological conditions, has been developed using thiol-Michael addition reaction. The mechanical properties, gelation process and degree of swelling of the hydrogel can be easily adjusted by changing the pH of phosphate buffer saline. The 3D cell encapsulation ability is demonstrated by encapsulating rat bone marrow mesenchymal stem cells (BMSCs) and NIH/3T3 fibroblasts into the in situ-forming hydrogel with maintained high viability. The BMSCs also maintain their differentiation potential after encapsulation. These results demonstrate that the Dex-l-DTT hydrogel holds great potential for biomedical field.