EMAIL THIS PAGE TO A FRIEND
Journal of diabetes and its complications
Decreased expression of heat shock proteins may lead to compromised wound healing in type 2 diabetes mellitus patients.
PMID 25746357
Abstract
Heat shock proteins (HSPs) are inducible stress proteins expressed in cells exposed to stress. HSPs promote wound healing by recruitment of dermal fibroblasts to the site of injury and bring about protein homeostasis. Diabetic wounds are hard to heal and inadequate HSPs may be important contributors in the etiology of diabetic foot ulcers (DFU). To analyze the differential expression of HSPs and their downstream molecules in human diabetic wounds compared to control wounds. Expressional levels of HSP27, HSP47 and HSP70 and their downstream molecules like TLR4, p38-MAPK were seen in biopsies from 101 human diabetic wounds compared to 8 control subjects without diabetes using RT-PCR, western blot and immunohistochemistry. Our study suggested a significant down regulation of HSP70, HSP47 and HSP27 (p value=<0.001 for HSP70; p value=0.007 for HSP47; p value=0.007 for HSP27) in DFU along with their downstream molecules TLR4 and p38-MAPK (p value=0.006 for p38-MAPK; p value=0.02 for TLR4). HSP70 levels were significantly lower in male subjects and their levels increased significantly with the grades of wound on Wagner's scale. Infection status of the wounds was found to be significantly associated with the increased levels of HSP70 and HSP27 in infected diabetic wounds. Our study demonstrates that the down regulation of HSPs in diabetic wounds is associated with wound healing impairment in T2DM subjects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1306362
ANTI-HSP27 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501456
Anti-HSP27 antibody produced in rabbit, affinity isolated antibody
SAB4501457
Anti-HSP27 antibody produced in rabbit, affinity isolated antibody
SAB4300495
Anti-HSPB1 (Ab-78) antibody produced in rabbit, affinity isolated antibody
SAB4300496
Anti-HSPB1 (Ab-82) antibody produced in rabbit, affinity isolated antibody
AV09060
Anti-HSPB1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV30177
Anti-HSPB1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA000497
Anti-HSPB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701965
Anti-HSPB1 antibody produced in rabbit
HPA038889
Anti-HSPB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB5200099 Anti-HSPB2 antibody produced in rabbit, 1 mg/mL, affinity isolated antibody
SAB4501458
Anti-HSPB2, C-Terminal antibody produced in rabbit, affinity isolated antibody
S5950
Anti-SERPINH1 (404-418) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1405565
Anti-SERPINH1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA029198
Anti-SERPINH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3701437
Monoclonal Anti-HSP27 antibody produced in mouse, clone G3.1, purified immunoglobulin
WH0000871M1
Monoclonal Anti-SERPINH1 antibody produced in mouse, clone 1D2-1A6, purified immunoglobulin, buffered aqueous solution