EMAIL THIS PAGE TO A FRIEND
FEBS letters
Msi1 confers resistance to TRAIL by activating ERK in liver cancer cells.
PMID 25747387
Abstract
To investigate TRAIL resistance mechanisms in hepatocellular carcinoma (HCC), we isolated a stable TRAIL-resistant sub-population of the HCC cell line LH86, designated LH86-TR. Differential activation of AKT was not responsible for acquisition of TRAIL resistance. Cells with both congenital and acquired resistance to TRAIL exhibited increased Msi1 expression, which conferred TRAIL resistance by activating ERK. Forced expression of Msi1 decreased the sensitivity of HCC cells to TRAIL both in vitro and in vivo. Conversely, shRNA-mediated depletion of Msi1 enhanced TRAIL efficacy. SiRNA-mediated depletion of ERK overcame TRAIL resistance. Hence, we conclude that Msi1 is a mediator of TRAIL resistance in HCC cells.