EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Regulation of store-operated Ca2+ entry activity by cell cycle dependent up-regulation of Orai2 in brain capillary endothelial cells.
PMID 25748572
Abstract
Store-operated Ca(2+) entry (SOCE) via Orai1 and STIM1 complex is supposed to have obligatory roles in the regulation of cellular functions of vascular endothelial cells, while little is known about the contribution of Orai2. Quantitative PCR and Western blot analyses indicated the expression of Orai2 and STIM2, in addition to Orai1 and STIM1 in bovine brain capillary endothelial cell line, t-BBEC117. During the exponential growth of t-BBEC117, the knockdown of Orai1 and STIM1 significantly reduced the SOCE activity, whereas Orai2 and STIM2 siRNAs had no effect. To examine whether endogenous SOCE activity contributes to the regulation of cell cycle progression, t-BBEC117 were synchronized using double thymidine blockage. At the G2/M phase, Ca(2+) influx via SOCE was decreased and Orai2 expression was increased compared to the G0/G1 phase. When Orai2 was knocked down at the G2/M phase, the decrease in SOCE was removed, and cell proliferation was partly attenuated. Taken together, Orai1 significantly contributes to cell proliferation via the functional expression, which is presumably independent of the cell cycle phases. In construct, Orai2 is specifically up-regulated during the G2/M phase, negatively modulates the SOCE activity, and may contribute to the regulation of cell cycle progression in brain capillary endothelial cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
439770
Gadolinium(III) chloride, anhydrous, powder, 99.99% trace metals basis
Cl3Gd
735949
Gadolinium(III) chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
Cl3Gd
612081
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), DSS 0.1 %, water 1 %, NMR tube size 3 mm × 8 in.
Cl3Gd
487139
Gadolinium(III) chloride solution, NMR reference standard, water 1 %, DSS 0.1 %, 0.1 mg/mL in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Cl3Gd
611948
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), DSS 0.1 %, water 1 %, NMR tube size 8 mm × 8 in.
Cl3Gd
721905
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), Methanol-13C 0.1 % (99 atom % 13C), water 1 %, NMR tube size 3 mm × 8 in.
Cl3Gd
721913
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), water 1 %, Methanol-13C 0.1 % (99 atom % 13C), NMR tube size 5 mm × 8 in.
Cl3Gd
703656
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), water 0.1 %, NMR tube size 6.5 mm × 8 in.
Cl3Gd
449830
Lanthanum(III) chloride, anhydrous, beads, −10 mesh, ≥99.99% trace metals basis
Cl3La
298182
Lanthanum(III) chloride, anhydrous, beads, −10 mesh, 99.9% trace metals basis
Cl3La