EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Pd-catalyzed P-arylation of triarylantimony dicarboxylates with dialkyl H-phosphites without a base: synthesis of arylphosphonates.
PMID 25748784
Abstract
The reaction of triarylantimony diacetates [Ar3Sb(OAc)2] with dialkyl H-phosphites [H-PO(OR)2] in the presence of a Pd(PPh3)4 (5 mol%) catalyst led to the formation of arylphosphonates in moderate to excellent yield under base-free conditions. This reaction is the first example of carbon-phosphorus bond formation by using an organoantimony compound as a pseudo-halide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
142166
Triphenylantimony(V) diacetate, 97%
C22H21O4Sb