EMAIL THIS PAGE TO A FRIEND
Acta biomaterialia
Enzymatic regulation of functional vascular networks using gelatin hydrogels.
PMID 25749296
Abstract
To manufacture tissue engineering-based functional tissues, scaffold materials that can be sufficiently vascularized to mimic the functionality and complexity of native tissues are needed. Currently, vascular network bioengineering is largely carried out using natural hydrogels as embedding scaffolds, but most natural hydrogels have poor mechanical stability and durability, factors that critically limit their widespread use. In this study, we examined the suitability of gelatin-phenolic hydroxyl (gelatin-Ph) hydrogels that can be enzymatically crosslinked, allowing tuning of the storage modulus and the proteolytic degradation rate, for use as injectable hydrogels to support the human progenitor cell-based formation of a stable and mature vascular network. Porcine gelatin-Ph hydrogels were found to be cytocompatible with human blood-derived endothelial colony-forming cells and white adipose tissue-derived mesenchymal stem cells, resulting in >87% viability, and cell proliferation and spreading could be modulated by using hydrogels with different proteolytic degradability and stiffness. In addition, gelatin was extracted from mouse dermis and murine gelatin-Ph hydrogels were prepared. Importantly, implantation of human cell-laden porcine or murine gelatin-Ph hydrogels into immunodeficient mice resulted in the rapid formation of functional anastomoses between the bioengineered human vascular network and the mouse vasculature. Furthermore, the degree of enzymatic crosslinking of the gelatin-Ph hydrogels could be used to modulate cell behavior and the extent of vascular network formation in vivo. Our report details a technique for the synthesis of gelatin-Ph hydrogels from allogeneic or xenogeneic dermal skin and suggests that these hydrogels can be used for biomedical applications that require the formation of microvascular networks, including the development of complex engineered tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8001
3,3′-Diaminobenzidine, ≥99% (HPLC)
C12H14N4
D12384
3,3′-Diaminobenzidine, 99%
C12H14N4
P9346
Liquified Phenol, meets USP testing specifications, ≥89.0%
C6H6O
M3671
MES, low moisture content, ≥99% (titration)
C6H13NO4S
76039
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
C6H13NO4S
P1037
Phenol, for molecular biology
C6H6O
P5566
Phenol, BioXtra, ≥99.5% (GC)
C6H6O
328111
Phenol, unstabilized, purified by redistillation, ≥99%
C6H6O
242322
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
C6H6O
77607
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
C6H6O
33517
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
C6H6O
16016
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
C6H6O
16017
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
C6H6O
W322318
Phenol, ≥99%
C6H6O
W322340
Phenol, natural, 97%, FG
C6H6O
P4557
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
C6H6O
P4682
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
C6H6O
T90344
Tyramine, 99%
C8H11NO
W421501
Tyramine, 98%, FG
C8H11NO