EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Analysis of perfluorooctanoic acid by high performance liquid chromatography with 3,4-dichloroaniline derivatization].
PMID 25752085
Abstract
A simple derivatization method followed by high performance liquid chromatography (HPLC) for the analysis of perfluorooctanoic acid (PFOA) was developed. PFOA was firstly derivatized with 3,4-dichloroaniline (DCA) using carbodiimide method. The typical amidate product was characterized by mass spectrometry (MS). It can be detected by a UV detector at maximum absorption wavelength of 255 nm. It was isolated well by methanol/H2O (8/2, v/v) as mobile phase. The DCA derivatization of PFOA in organic or aqueous solutions was opti- mized. The following purification procedures with thin layer chromatography (TLC) and HPLC separations were established. Using external standard method with the authentic PFOA-DCA derivative as standard, the limit of instrumental quantification was found to be 0. 5 mg/L. Good linear correlation coefficients were observed in 0.5-50.0 mg/L. The accuracy of the method was evaluated by the recovery measurements on spiked samples. The recoveries for the spiked samples (1.0 mg/L) in organic phase and aqueous phase were 91.8%-108.7% and 42.1%-53.7%, respectively. Comparing to the reported pre-column derivatization methods followed by HPLC for the determination of PFOA, this method has the advantages of mild reaction conditions, very stable derivative, easy operation, low cost, etc. The method was successfully applied for the quantification of PFOA in photodegradation experiments, and the results were consistent with those determined by LC/MS. Considering the combination of the preconcentration step, it has a potential for the application of the analysis of samples containing relatively low concentrations, such as μg/L levels of PFOA obtained from environmental or scientific experiments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
437778
3,4-Dichloroaniline, 98%
C6H5Cl2N