EMAIL THIS PAGE TO A FRIEND
Journal of cellular physiology
Cross-Talk Between VEGF and BMP-6 Pathways Accelerates Osteogenic Differentiation of Human Adipose-Derived Stem Cells.
PMID 25753222
Abstract
Deficiency in vascular endothelial growth factor (VEGF) or bone morphogenetic proteins (BMPs) results in fracture non-unions. Therefore, it is indispensable to comprehend the combined effect of VEGF and BMPs on the osteogenic differentiation of osteoprogenitor mesenchymal stem cells (MSCs) that are either naturally occurring at the fracture repair site or exogenously added to enhance the bone repair. We found that the combination of VEGF and BMP-6 enhanced COL1A2 expression, which correlated with upregulated expression of osterix, Dlx5, and Msx2 in human adipose-derived stem cells (hADSCs). Cross-talk between VEGF and BMP-6 pathways upregulated activation of p38 mitogen-activated kinase (p38 MAPK) and inhibited activation of protein kinase B (PKB, also known as Akt), whereas phosphorylation of "mothers against decapentaplegic" homologs 1/5/8 (Smads 1/5/8) and extracellular signal-regulated kinases 1 and 2 (ERK 1/2) was not affected. Consistent with these findings, p38 inhibitor SB203580, or siRNA knockdown of osterix, abrogated crosstalk between the VEGF and BMP-6 pathways and significantly reduced the observed upregulation of COL1A2. Nuclear translocation of the phosphorylated form of osterix was also inhibited by SB203580. Although crosstalk between the VEGF-BMP-6 pathways did not show an effect on the extent of mineralization, inhibition of any one of the three components that were upregulated through the cross-talk, i.e., osterix, Dlx5, and p38 activation, led to a complete inhibition of mineralization. Inhibition of PKB/Akt activation, which is attenuated through the cross-talk, significantly enhanced ALP gene expression. These observations imply that crosstalk between the VEGF and BMP-6 signaling pathways enhances osteogenic differentiation of MSCs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100463
Anti-COL1A2 antibody produced in rabbit, affinity isolated antibody
AV31604
Anti-DLX2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV32212
Anti-DLX2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA039071
Anti-DLX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV38586
Anti-DLX5 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV38585
Anti-DLX5 (AB2) antibody produced in rabbit, IgG fraction of antiserum
GW22362A Anti-DLX5 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501236
Anti-DLX5 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1412173
ANTI-DLX5 antibody produced in mouse, clone 4F5, purified immunoglobulin, buffered aqueous solution
SAB1412288
ANTI-DLX5 antibody produced in mouse, clone 4C6, purified immunoglobulin, buffered aqueous solution
SAB1412489
ANTI-DLX5 antibody produced in mouse, clone 1B7, purified immunoglobulin, buffered aqueous solution
SAB1412490 ANTI-DLX5 antibody produced in mouse, clone 3F5, purified immunoglobulin, buffered aqueous solution
SAB1412491
ANTI-DLX5 antibody produced in mouse, clone 4H6, purified immunoglobulin, buffered aqueous solution
SAB1412492
ANTI-DLX5 antibody produced in mouse, clone 2B6, purified immunoglobulin, buffered aqueous solution
SAB1412664
ANTI-DLX5 antibody produced in mouse, clone 4B7, purified immunoglobulin, buffered aqueous solution
HPA005670
Anti-DLX5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32867
Anti-DLX5 antibody produced in rabbit, affinity isolated antibody
SAB4500660
Anti-DLX5 antibody produced in rabbit, affinity isolated antibody
SAB2103442
Anti-DLX5, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4504206
Anti-phospho-Smad1 (pSer187) antibody produced in rabbit, affinity isolated antibody
SAB4503779
Anti-phospho-Smad1 (pSer465) antibody produced in rabbit, affinity isolated antibody
SAB4300250
Anti-phospho-SMAD1 (pSer465) antibody produced in rabbit, affinity isolated antibody
AV31622
Anti-SP7 antibody produced in rabbit, affinity isolated antibody
SAB2104296
Anti-SP7, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1402731
Monoclonal Anti-COL1A2, (C-terminal) antibody produced in mouse, clone 7E11, purified immunoglobulin, buffered aqueous solution
WH0001746M1
Monoclonal Anti-DLX2 antibody produced in mouse, clone 2H2, purified immunoglobulin, buffered aqueous solution
SAB1408949
Monoclonal Anti-DLX2 antibody produced in mouse, clone 3H3, purified immunoglobulin, buffered aqueous solution
SAB1408950
Monoclonal Anti-DLX2 antibody produced in mouse, clone 3G6, purified immunoglobulin, buffered aqueous solution
SAB1408951
Monoclonal Anti-DLX2 antibody produced in mouse, clone 1E9, purified immunoglobulin, buffered aqueous solution
SAB1403739
Monoclonal Anti-DLX2, (N-terminal) antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution