EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Involvement of ATM in homologous recombination after end resection and RAD51 nucleofilament formation.
PMID 25753674
Abstract
Ataxia-telangiectasia mutated (ATM) is needed for the initiation of the double-strand break (DSB) repair by homologous recombination (HR). ATM triggers DSB end resection by stimulating the nucleolytic activity of CtIP and MRE11 to generate 3'-ssDNA overhangs, followed by RPA loading and RAD51 nucleofilament formation. Here we show for the first time that ATM is also needed for later steps in HR after RAD51 nucleofilament formation. Inhibition of ATM after completion of end resection did not affect RAD51 nucleofilament formation, but resulted in HR deficiency as evidenced by (i) an increase in the number of residual RAD51/γH2AX foci in both S and G2 cells, (ii) the decrease in HR efficiency as detected by HR repair substrate (pGC), (iii) a reduced SCE rate and (iv) the radiosensitization of cells by PARP inhibition. This newly described role for ATM was found to be dispensable in heterochromatin-associated DSB repair, as KAP1-depletion did not alleviate the HR-deficiency when ATM was inhibited after end resection. Moreover, we demonstrated that ATR can partly compensate for the deficiency in early, but not in later, steps of HR upon ATM inhibition. Taken together, we describe here for the first time that ATM is needed not only for the initiation but also for the completion of HR.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501369
Anti-Histone H2AX , C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1406364
Anti-RAD51 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101936
Anti-RAD51 antibody produced in rabbit, affinity isolated antibody
SAB2700175
Anti-RAD51 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA039310
Anti-RAD51 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502565
Anti-RAD51, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502566
Anti-RAD51, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1406602
Anti-XRCC2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
X3878 Anti-XRCC2 antibody produced in rabbit, affinity isolated antibody, ammonium sulfate suspension
SAB2102727
Anti-XRCC2 antibody produced in rabbit, affinity isolated antibody
SAB1411401
Anti-XRCC2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
EHU091361 MISSION® esiRNA, esiRNA human MRE11A (esiRNA1)
EMU012341 MISSION® esiRNA, esiRNA targeting mouse Mre11a (esiRNA1)
EHU067331 MISSION® esiRNA, esiRNA human TRIM28 (esiRNA1)
EMU024081 MISSION® esiRNA, esiRNA targeting mouse Trim28 (esiRNA1)
EHU060331 MISSION® esiRNA, esiRNA human CDKN3 (esiRNA1)
EMU165011 MISSION® esiRNA, esiRNA targeting mouse Cdkn3
SIHP0064 MISSION® Human Phosphatase, CDKN3 siRNA1, Nano Scale 250 pmol
SIHP0065 MISSION® Human Phosphatase, CDKN3 siRNA2, Nano Scale 250 pmol
SIHP0066 MISSION® Human Phosphatase, CDKN3 siRNA3, Nano Scale 250 pmol
WH0005888M1
Monoclonal Anti-RAD51 antibody produced in mouse, clone 2E5-E5, purified immunoglobulin, buffered aqueous solution
SAB1404293
Monoclonal Anti-RAD51 antibody produced in mouse, clone 4C11-B4, purified immunoglobulin, buffered aqueous solution
SAB1402394
Monoclonal Anti-XRCC2 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution