EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Preparation and characterization of amphiphilic calixarene nanoparticles as delivery carriers for paclitaxel.
PMID 25757488
Abstract
Two types of amphoteric calix[n]arene carboxylic acid (CnCA) derivative, i.e., calix[6]arene hexa-carboxylic acid (C6HCA) and calix[8]arene octo-carboxylic acid (C8OCA), were synthesized by introducing acetoxyls into the hydroxyls of calix[n]arene (n=6, 8). C6HCA and C8OCA nanoparticles (NPs) were prepared successfully using the dialysis method. CnCA NPs had regular spherical shapes with an average diameter of 180-220 nm and possessed negative charges of greater than -30 mV. C6HCA and C8OCA NPs were stable in 4.5% bovine serum albumin solutions and buffers (pH 5-9), with a low critical aggregation concentration value of 5.7 mg·L(-1) and 4.0 mg·L(-1), respectively. C6HCA and C8OCA NPs exhibited good paclitaxel (PTX) loading capacity, with drug loading contents of 7.5% and 8.3%, respectively. The overall in vitro release behavior of PTX from the CnCA NPs was sustained, and C8OCA NPs had a slower release rate compared with C6HCA NPs. These favorable properties of CnCA NPs make them promising nanocarriers for tumor-targeted drug delivery.