EMAIL THIS PAGE TO A FRIEND
Applied microbiology and biotechnology
Optimization of recombinant expression enables discovery of novel cytochrome P450 activity in rice diterpenoid biosynthesis.
PMID 25758958
Abstract
The oxygenation reactions catalyzed by cytochromes P450 (CYPs) play critical roles in plant natural products biosynthesis. At the same time, CYPs are one of most challenging enzymes to functionally characterize due to the difficulty of recombinantly expressing these membrane-associated monooxygenases. In the course of investigating rice diterpenoid biosynthesis, we have developed a synthetic biology approach for functional expression of relevant CYPs in Escherichia coli. In certain cases, activity was observed for only one of two closely related paralogs although it seems clear that related reactions are required for production of the known diterpenoids. Here, we report that optimization of the recombinant expression system enabled characterization of not only these previously recalcitrant CYPs, but also discovery of additional activity relevant to rice diterpenoid biosynthesis. Of particular interest, CYP701A8 was found to catalyze 3β-hydroxylation of syn-pimaradiene, which is presumably relevant to momilactone biosynthesis, while CYP71Z6 & 7 were found to catalyze multiple reactions, with CYP71Z6 catalyzing the production of 2α,3α-dihydroxy-ent-isokaurene via 2α-hydroxy-ent-isokaurene, and CYP71Z7 catalyzing the production of 3α-hydroxy-ent-cassadien-2-one via 2α-hydroxy-ent-cassadiene and ent-cassadien-2-one, which may be relevant to oryzadione and phytocassane biosynthesis, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0440
Spautin-1, ≥98% (HPLC)
C15H11F2N3