EMAIL THIS PAGE TO A FRIEND
PLoS genetics
Effects on murine behavior and lifespan of selectively decreasing expression of mutant huntingtin allele by supt4h knockdown.
PMID 25760041
Abstract
Production of protein containing lengthy stretches of polyglutamine encoded by multiple repeats of the trinucleotide CAG is a hallmark of Huntington's disease (HD) and of a variety of other inherited degenerative neurological and neuromuscular disorders. Earlier work has shown that interference with production of the transcription elongation protein SUPT4H results in decreased cellular capacity to transcribe mutant huntingtin gene (Htt) alleles containing long CAG expansions, but has little effect on expression of genes containing short CAG stretches. zQ175 and R6/2 are genetically engineered mouse strains whose genomes contain human HTT alleles that include greatly expanded CAG repeats and which are used as animal models for HD. Here we show that reduction of SUPT4H expression in brains of zQ175 mice by intracerebroventricular bolus injection of antisense 2'-O-methoxyethyl oligonucleotides (ASOs) directed against Supt4h, or in R6/2 mice by deletion of one copy of the Supt4h gene, results in a decrease in mRNA and protein encoded specifically by mutant Htt alleles. We further show that reduction of SUPT4H in mouse brains is associated with decreased HTT protein aggregation, and in R6/2 mice, also with prolonged lifespan and delay of the motor impairment that normally develops in these animals. Our findings support the view that targeting of SUPT4H function may be useful as a therapeutic countermeasure against HD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32883
4-tert-Octylphenol monoethoxylate solution, 1 μg/mL in acetone, analytical standard
C16H26O2
32882
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
C16H26O2
MAB2166
Anti-Huntingtin Protein Antibody, a.a. 181-810, clone 1HU-4C8, 100 µL, Anti-Huntingtin Protein Antibody, a.a. 181-810, clone 1HU-4C8 is an antibody against Huntingtin Protein for use in ELISA, IC, IH(P), IP & WB.
MAB5374
Anti-Huntingtin Protein Antibody, clone mEM48, 100 µL, Detect Huntingtin Protein using this Anti-Huntingtin Protein Antibody, clone mEM48 validated for use in IC, IH & WB.
MAB1574
Anti-Polyglutamine-Expansion Diseases Marker Antibody, clone 5TF1-1C2, 100 µL, Anti-Polyglutamine-Expansion Diseases Marker Antibody, clone 5TF1-1C2 is an antibody against Polyglutamine-Expansion Diseases Marker for use in ELISA, IC, IH(P), IP & WB.
1012772
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
C2H6O
PHR1070
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
C2H6O
47072
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
FNa
80373
Fluoride, certified anion standard solution, 1000 mg/kg F-, for ion chromatography, certified by BAM, according to ISO 17025
FNa
PHG0010
Isopropyl β-D-1-thiogalactopyranoside, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
C9H18O5S
T1827
Monoclonal Anti-TBP antibody produced in mouse, ~2.0 mg/mL, clone 58C9, purified immunoglobulin, buffered aqueous solution
95660
o-Xylene, analytical standard
C8H10
294780
o-Xylene, anhydrous, 97%
C8H10
295884
o-Xylene, for HPLC, 98%
C8H10
X1040
o-Xylene, reagent grade, ≥98.0%
C8H10
PHR1312
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10
71383
Sodium chloride, tested according to Ph.Eur.
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
ClNa
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa
450022
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
FNa
PHR1408
Sodium Fluoride, certified reference material
FNa
67414
Sodium fluoride 0.5 M solution
FNa
342483
Sodium phosphate, 96%
Na3O4P
71177
Sodium standard for AAS, ready-to-use, traceable to BAM, in H2O
ClNa
T7567
Tributylphosphine solution, 200 mM (in N-methyl-2-pyrrolidinone), liquid
C12H27P