EMAIL THIS PAGE TO A FRIEND
Oncotarget
Deficiency of AMPK in CD8+ T cells suppresses their anti-tumor function by inducing protein phosphatase-mediated cell death.
PMID 25760243
Abstract
A number of studies have linked AMPK, a major metabolic sensor coordinating of multiple cellular functions, to tumor development and progression. However, the exact role of AMPK in tumor development is still controversial. Here we report that activation of AMPK promotes survival and anti-tumor function of T cells, in particular CD8+ T cells, resulting in superior tumor suppression in vivo. While AMPK expression is dispensable for T cell development, genetic deletion of AMPK promotes T cell death during in vitro activation and in vivo tumor development. Moreover, we demonstrate that protein phosphatases are the key mediators of AMPK-dependent effects on T cell death, and inhibition of phosphatase activity by okadaic acid successfully restores T cell survival and function. Altogether, our data suggest a novel mechanism by which AMPK regulates protein phosphatase activity in control of survival and function of CD8+ T cells, thereby enhancing their role in tumor immunosurveillance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
20021
3-tert-Butyl-4-hydroxyanisole, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)
C11H16O2