EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Viscosupplementation for osteoarthritis of the knee.
PMID 25760356
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7630
Hyaluronic acid sodium salt from bovine vitreous humor
H1876
Hyaluronic acid sodium salt from human umbilical cord
H5388
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
53747
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
49775
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
40583
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
94137
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
97616
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
42686
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
08185
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000
75048
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-90,000
96144
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-120,000
75046
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 90,000-110,000
06742
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 120,000-350,000
75043
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 130,000-150,000
75044
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 150,000-300,000
00941
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
75574
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 500,000-750,000
53163
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 750,000-1,000,000
41897
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
80486
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,250,000-1,500,000
63357
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
02737
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,750,000-2,000,000
51967
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
73641
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
H9390
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide